Mesalamine
Synonym(s):
5-Aminosalicylic acid, 5-AS, 5-Amino-2-hydroxybenzoic acid, Mesalamine
Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing