Skip to Content
Merck
G-418 Solution solution, =98% (TLC), suitable for transfection

G-418 Solution

G-418 Solution solution, =98% (TLC), suitable for transfection
Synonym(s):
G 418 disulfate salt solution, Antibiotic G418
Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71

Products

Product Category
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing