Calcium chloride solution
Synonym(s):
Calcium chloride solution
Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Format
Quality Segment
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing