Skip to Content
MY
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
fritted disc
Synonym(s):
Corning
®
33950
Products
Manufacturer
Diameter (mm)
Pore size (µm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS33950MEC
Pyrex
®
fritted disc,
body diam. 35 mm, porosity 170-220 μm
Expand
View Pricing
CLS33950SC
Pyrex
®
fritted disc,
body diam. 25 mm, porosity 40-60 μm
Expand
View Pricing
CLS33950LEC
Pyrex
®
fritted disc,
body diam. 45 mm, porosity 170-220 μm
Expand
View Pricing
CLS33950LC
Pyrex
®
fritted disc,
body diam. 45 mm, porosity 40-60 μm
Expand
View Pricing
CLS33950MC
Pyrex
®
fritted disc,
body diam. 35 mm, porosity 40-60 μm
Expand
View Pricing