Skip to Content
MY
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium bicarbonate
Synonym(s):
Ammonium hydrogen carbonate
Linear Formula:
NH
4
HCO
3
CAS Number:
1066-33-7
Molecular Weight:
79.06
EC Number:
213-911-5
Products
Product Category
salts (3)
buffers (1)
Brand
SAFC (1)
Sigma-Aldrich (4)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Puriss (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
A6141
Ammonium bicarbonate,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
09830
Ammonium bicarbonate,
BioUltra, ≥99.5% (T)
Expand
View Pricing
11213
Ammonium bicarbonate,
puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%
Expand
View Pricing
1.01131
Ammonium bicarbonate,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, E 503
Expand
View Pricing
V800035
Ammonium bicarbonate,
LR, ≥98.5%
Expand
View Pricing