Skip to Content
Merck
Salbutamol hemisulfate salt ≥98%

Salbutamol hemisulfate salt

Salbutamol hemisulfate salt ≥98%
Synonym(s):
α-([t-Butylamino]methyl)-4-hydroxy-m-xylene-α,α′-diol, Albuterol
Linear Formula:
C13H21NO3 · 1/2H2SO4
CAS Number:
Molecular Weight:
288.35
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing