Skip to Content
Merck
2,6-Diisopropylphenyl isocyanate technical grade, 90%

2,6-Diisopropylphenyl isocyanate

2,6-Diisopropylphenyl isocyanate technical grade, 90%
Linear Formula:
[(CH3)2CH]2C6H3NCO
CAS Number:
Molecular Weight:
203.28
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing