Skip to Content
Merck
MOPS Vetec™, reagent grade

MOPS

MOPS Vetec™, reagent grade
Synonym(s):
3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid
Empirical Formula (Hill Notation):
C7H15NO4S
CAS Number:
Molecular Weight:
209.26
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Technique