Skip to Content
Merck
N-Methyl-N-trimethylsilyltrifluoroacetamide activated III for GC derivatization, LiChropur™, activated with imidazole

N-Methyl-N-(trimethylsilyl)trifluoroacetamide

N-Methyl-N-trimethylsilyltrifluoroacetamide activated III for GC derivatization, LiChropur™, activated with imidazole
Synonym(s):
N-Trimethylsilyl-N-methyl trifluoroacetamide, MSTFA, N-Methyl-N-trimethylsilyltrifluoroacetamide imidazole mixture, MSTFA activated III
Linear Formula:
CF3CON(CH3)Si(CH3)3
CAS Number:
Molecular Weight:
199.25