Merck
Pesticides by US EPA Method IP-8, TO-10a, ASTM® D 4861
Pesticides by US EPA Method IP-8, TO-10a, ASTM® D 4861 suitable for GC, application for air monitoring
Materiales
analytical column
Referencia del producto
Descripción
Precios
SPB®-608
L × I.D. 30 m × 0.53 mm, df 0.50
Related product
Referencia del producto
Descripción
Precios
ORBO 1000 Precleaned Small PUF Cartridge
O.D. × L 22 mm × 7.6 cm, pkg of 3 ea
CONDITIONS
sample/matrix
polyurethane foam (PUF) spiked with 250 ng each pesticide
adsorbent tube
ORBO-1000 (20557)
column
SPB-608, 30 m × 0.53 mm I.D., 0.5 μm film (25312)
oven
150 °C (1 min) to 270 °C at 4 °C/min
detector
ECD
carrier gas
helium, 7 mL/min
injection
1 μL
Descripción
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany