Key Documents
TMO312104-0002
Sigma-Aldrich
Nalgene® wide-mouth bottle
high-density polyethylene, capacity 60 mL, natural, case of 1000 ea
About This Item
UNSPSC Code:
41121800
material
high-density polyethylene
description
agency approvals (Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.)
packaging
case of 1000 ea
manufacturer/tradename
Nalge-Nunc 312104-0002
capacity
60 mL
color
natural
General description
Confidently ship and store these Thermo Scientific Nalgene Wide-Mouth HDPE Packaging Bottles, which are easy to fill and resistant to chemicals. These wide-mouthed, high-density polyethelyene bottles are injection blow-molded for strength and uniform wall thickness.
Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries