Merck
Attenuation of the Infiltration of Angiotensin II Expressing CD3
Frontiers in molecular neuroscience (2017-12-05)
Nemat Khan, Arjun Muralidharan, Maree T Smith
ABSTRACT
Recent preclinical and proof-of-concept clinical studies have shown promising analgesic efficacy of selective small molecule angiotensin II type 2 (AT