- Clinical significance of platelet-derived growth factor receptor-β gene expression in stage II/III gastric cancer with S-1 adjuvant chemotherapy.
Clinical significance of platelet-derived growth factor receptor-β gene expression in stage II/III gastric cancer with S-1 adjuvant chemotherapy.
Oncology letters (2017-03-31)
Akio Higuchi, Takashi Oshima, Kazue Yoshihara, Kentaro Sakamaki, Toru Aoyama, Nobuyasu Suganuma, Naoto Yamamoto, Tsutomu Sato, Haruhiko Cho, Manabu Shiozawa, Takaki Yoshikawa, Yasushi Rino, Chikara Kunisaki, Toshio Imada, Munetaka Masuda
PMID28356977
ABSTRACT
Overall survival remains unsatisfactory in stage II/III gastric cancer, even after curative surgery and adjuvant chemotherapy. Platelet-derived growth factor receptor-β (