Merck
  • Quantitation of human haptoglobin: comparative ELISA studies using adsorption and capture methods.
Quantitation of human haptoglobin: comparative ELISA studies using adsorption and capture methods.
Journal of immunological methods (1999-06-12)
E Yee, J E Brown
ABSTRACT
Three ELISA methods for the quantitation of haptoglobin (Hp) in plasma and albumin are described: a polystyrene direct adsorption method and capture methods with antibody and hemoglobin. Hp aggregates generated by 60 degrees C heating showed as much as a hundred-fold higher response by polystyrene adsorption compared to the two capture methods, while unheated Hp showed comparable responses by the three methods.