Merck
  • New whole-body multimodality imaging of gastric cancer peritoneal metastasis combining fluorescence imaging with ICG-labeled antibody and MRI in mice.
New whole-body multimodality imaging of gastric cancer peritoneal metastasis combining fluorescence imaging with ICG-labeled antibody and MRI in mice.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association (2013-11-30)
Akihiro Ito, Yuichi Ito, Shigeru Matsushima, Daisuke Tsuchida, Mai Ogasawara, Junichi Hasegawa, Kazunari Misawa, Eisaku Kondo, Norio Kaneda, Hayao Nakanishi
ABSTRACT
Peritoneal metastasis is the most frequent pattern of recurrence after curative surgery for gastric cancer. However, such a recurrence is difficult to detect by conventional computed tomography (CT) and magnetic resonance imaging (MRI) at an early stage. To improve the sensitivity and specificity of diagnostic imaging for peritoneal metastasis, we developed a new type of multimodality imaging combining fluorescence imaging with near-infrared fluorophore (NIR)-labeled antibodies and MRI. Dual optical imaging of peritoneal metastasis was carried out using luciferase-tagged gastric cancer cell lines and XenoLight CF750 or indocyanine green (ICG)-labeled anti-human epidermal growth factor receptor (EGFR) or CEA antibody as a probe in mice with Ivis in vivo imaging system. This whole-body fluorescent imaging system sensitively detected metastatic foci <1 mm in diameter in the peritoneal cavity noninvasively. Fluorescence imaging proved to be specific because the fluorescence signal was abolished by blocking with excess unlabeled antibody. Although this fluorescence imaging had higher sensitivity for detection of small-sized peritoneal metastases than MRI, it proved difficult to accurately determine organ distribution of the metastasis. We thus developed a multimodality imaging system by the fusion of the three-dimensional fluorescence image with the MRI image and demonstrated its improved diagnostic accuracy over either method alone. The present results suggest that multimodality imaging consisting of fluorescence imaging with NIR-labeled EGFR or CEA antibody and MRI allows sensitive, specific, and anatomically accurate detection of peritoneal metastasis noninvasively at an early stage.