Merck
  • Comparison of non-vertebral fracture between minodronate and risedronate therapy in elderly female patients with Alzheimer disease.
Comparison of non-vertebral fracture between minodronate and risedronate therapy in elderly female patients with Alzheimer disease.
Journal of musculoskeletal & neuronal interactions (2013-08-31)
Y Sato, Y Honda, J Iwamoto, N Amano
ABSTRACT
Minodronate is a nitrogen-containing bisphosphonate that is commercially available for the treatment of osteoporosis in Japan. Preclinical studies demonstrated that minodronate is at least 10 times more potent than alendronate in inhibiting bone resorption in vivo. A high incidence of fractures, particularly of the hip, represents an important problem in Alzheimer disease (AD) patients who are prone to falls and may have osteoporosis. A total of 256 elderly patients with AD were assigned to daily treatment with 1.0 mg of minodronate or a daily treatment with risedronate combined with daily 1000 IU ergocalciferol and 1200 mg elemental calcium, and followed up for 12 months. At baseline, patients of both groups showed low 25-hydroxyvitamin D with compensatory hyperparathyroidism. Non-vertebral fractures occurred in 5 patients in the minodronate group and 7 patients in the risedronate group (5 hip fractures; one fracture each at the distal forearm and pelvis). There was no difference in risk of hip fracture between the two groups (p=.70; odds ratio=0.8). The study medications were well tolerated with relatively few adverse events and were equivalent in reducing the risk of a fracture in elderly patients with AD.
Sigma-Aldrich
Risedronate sodium, ≥97% (HPLC)