Merck
  • Volatile constituents and antioxidant activity of flowers, stems and leaves of Nasturtium officinale R. Br.
Volatile constituents and antioxidant activity of flowers, stems and leaves of Nasturtium officinale R. Br.
Natural product research (2011-08-06)
Hamzeh Amiri
ABSTRACT
GC-MS analyses of the essential oils of leaves, stems and flower of Nasturtium officinale resulted in the identification of 9, 8 and 15 compounds, representing 97%, 100% and 94.7% of the oils, respectively. The main compounds of the oil of leaves were myristicin (57.6%), α-terpinolene (8.9%) and limonene (6.7%). Caryophyllene oxide (37.2%), p-cymene-8-ol (17.6%), α-terpinolene (15.2%) and limonene (11.8%) were the main components in stems, whereas limonene (43.6%), α-terpinolene (19.7%), p-cymene-8-ol (7.6%) and caryophyllene oxide (6.7%) were the major constituents in the oil of flowers. All the samples were subjected to a screening for their possible antioxidant activities using 2,2-diphenyl-1-picrylhydrazyl (DPPH) and β-carotene-linoleic acid assays. In the above tests, methanol extracts of leaves showed higher antioxidant activity than the oils and methanol extracts of stems and flowers.
Product Description
Supelco
Myristicin, analytical standard