Merck
  • FADD and caspase-8 mediate priming and activation of the canonical and noncanonical Nlrp3 inflammasomes.
FADD and caspase-8 mediate priming and activation of the canonical and noncanonical Nlrp3 inflammasomes.
Journal of immunology (Baltimore, Md. : 1950) (2014-01-24)
Prajwal Gurung, Paras K Anand, R K Subbarao Malireddi, Lieselotte Vande Walle, Nina Van Opdenbosch, Christopher P Dillon, Ricardo Weinlich, Douglas R Green, Mohamed Lamkanfi, Thirumala-Devi Kanneganti
ABSTRACT
The Nlrp3 inflammasome is critical for host immunity, but the mechanisms controlling its activation are enigmatic. In this study, we show that loss of FADD or caspase-8 in a RIP3-deficient background, but not RIP3 deficiency alone, hampered transcriptional priming and posttranslational activation of the canonical and noncanonical Nlrp3 inflammasome. Deletion of caspase-8 in the presence or absence of RIP3 inhibited caspase-1 and caspase-11 activation by Nlrp3 stimuli but not the Nlrc4 inflammasome. In addition, FADD deletion prevented caspase-8 maturation, positioning FADD upstream of caspase-8. Consequently, FADD- and caspase-8-deficient mice had impaired IL-1β production when challenged with LPS or infected with the enteropathogen Citrobacter rodentium. Thus, our results reveal FADD and caspase-8 as apical mediators of canonical and noncanonical Nlrp3 inflammasome priming and activation.