- YLZ-F5, a novel polo-like kinase 4 inhibitor, inhibits human ovarian cancer cell growth by inducing apoptosis and mitotic defects.
YLZ-F5, a novel polo-like kinase 4 inhibitor, inhibits human ovarian cancer cell growth by inducing apoptosis and mitotic defects.
Polo-like kinase 4 (PLK4), a member of the polo-like kinase family, plays several important roles in mitotic regulation, including centrosome duplication, spindle formation, and cytokinesis. PLK4 overexpression is frequently detected in many human cancers, including ovarian cancer, and the inhibition of PLK4 activity results in cancer cell mitotic arrest and apoptosis. Therefore, PLK4 might be a valid therapeutic target for antitumor therapy. In the present study, we aimed to determine if YLZ-F5, a potent small-molecule inhibitor of PLK4, inhibits ovarian cancer cell growth. MTT assay showed that YLZ-F5 inhibited ovarian cancer cell proliferation in a concentration- and time-dependent manner. The results of colony formation assays were consistent with those of the MTT assay results. In addition, YLZ-F5 induced ovarian cancer cell apoptosis that was associated with activation of caspase-3/caspase-9. Moreover, YLZ-F5 caused aberrant in centriole duplication that was associated with the inhibition of PLK4 phosphorylation. Notably, we showed that YLZ-F5 promoted the accumulation of ovarian cancer cells with mitotic defects (> 4 N DNA content) in a concentration-dependent manner. Furthermore, YLZ-F5 markedly inhibited the migration of A2780 cells. Taken together, these findings suggest that YLZ-F5 is a potential drug candidate for human ovarian cancer.