Merck
  • Strategy for the Biosynthesis of Short Oligopeptides: Green and Sustainable Chemistry.
Strategy for the Biosynthesis of Short Oligopeptides: Green and Sustainable Chemistry.
Biomolecules (2019-11-27)
Tao Wang, Yu-Ran Zhang, Xiao-Huan Liu, Shun Ge, You-Shuang Zhu
ABSTRACT
Short oligopeptides are some of the most promising and functionally important amide bond-containing components, with widespread applications. Biosynthesis of these oligopeptides may potentially become the ultimate strategy because it has better cost efficiency and environmental-friendliness than conventional solid phase peptide synthesis and chemo-enzymatic synthesis. To successfully apply this strategy for the biosynthesis of structurally diverse amide bond-containing components, the identification and selection of specific biocatalysts is extremely important. Given that perspective, this review focuses on the current knowledge about the typical enzymes that might be potentially used for the synthesis of short oligopeptides. Moreover, novel enzymatic methods of producing desired peptides via metabolic engineering are highlighted. It is believed that this review will be helpful for technological innovation in the production of desired peptides.
Sigma-Aldrich
Asp-Phe, 96%