Merck
  • Controlled protein release from monodisperse biodegradable double-wall microspheres of controllable shell thickness.
Controlled protein release from monodisperse biodegradable double-wall microspheres of controllable shell thickness.
Journal of controlled release : official journal of the Controlled Release Society (2013-08-21)
Yujie Xia, Pedro F Ribeiro, Daniel W Pack
ABSTRACT
Biodegradable polymer microparticles are promising delivery depots for protein therapeutics due to their relatively simple fabrication and facile administration. Double-wall microspheres (DWMS) comprising a core and shell made of two distinct polymers may provide enhanced control of the drug release profiles. Using precision particle fabrication (PPF) technology, monodisperse DWMS were fabricated with model protein bovine serum albumin (BSA)-loaded poly(lactide-co-glycolide) (PLG) core and drug-free poly(d,l-lactic acid) (PDLL) shell of uniform thickness. Monolithic single-wall microspheres were also fabricated to mimic the BSA-loaded PLG core. Using ethyl acetate and dichloromethane as shell- and core-phase solvents, respectively, BSA was encapsulated selectively in the core region within DWMS with higher loading and encapsulation efficiency compared to using dichloromethane as core and shell solvents. BSA in vitro release rates were retarded by the presence of the drug-free PDLL shell. Moreover, increasing PDLL shell thickness resulted in decreasing BSA release rate. With a 14-μm thick PDLL shell, an extended period of constant-rate release was achieved.
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
GYY4137 Dichloromethane complex, ≥98% (HPLC)