Merck
  • Angiopoietin 2 mediates microvascular and hemodynamic alterations in sepsis.
Angiopoietin 2 mediates microvascular and hemodynamic alterations in sepsis.
The Journal of clinical investigation (2013-07-19)
Tilman Ziegler, Jan Horstkotte, Claudia Schwab, Vanessa Pfetsch, Karolina Weinmann, Steffen Dietzel, Ina Rohwedder, Rabea Hinkel, Lisa Gross, Seungmin Lee, Junhao Hu, Oliver Soehnlein, Wolfgang M Franz, Markus Sperandio, Ulrich Pohl, Markus Thomas, Christian Weber, Hellmut G Augustin, Reinhard Fässler, Urban Deutsch, Christian Kupatt
ABSTRACT
Septic shock is characterized by increased vascular permeability and hypotension despite increased cardiac output. Numerous vasoactive cytokines are upregulated during sepsis, including angiopoietin 2 (ANG2), which increases vascular permeability. Here we report that mice engineered to inducibly overexpress ANG2 in the endothelium developed sepsis-like hemodynamic alterations, including systemic hypotension, increased cardiac output, and dilatory cardiomyopathy. Conversely, mice with cardiomyocyte-restricted ANG2 overexpression failed to develop hemodynamic alterations. Interestingly, the hemodynamic alterations associated with endothelial-specific overexpression of ANG2 and the loss of capillary-associated pericytes were reversed by intravenous injections of adeno-associated viruses (AAVs) transducing cDNA for angiopoietin 1, a TIE2 ligand that antagonizes ANG2, or AAVs encoding PDGFB, a chemoattractant for pericytes. To confirm the role of ANG2 in sepsis, we i.p. injected LPS into C57BL/6J mice, which rapidly developed hypotension, acute pericyte loss, and increased vascular permeability. Importantly, ANG2 antibody treatment attenuated LPS-induced hemodynamic alterations and reduced the mortality rate at 36 hours from 95% to 61%. These data indicate that ANG2-mediated microvascular disintegration contributes to septic shock and that inhibition of the ANG2/TIE2 interaction during sepsis is a potential therapeutic target.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction