Merck
  • Nigrostriatal overabundance of α-synuclein leads to decreased vesicle density and deficits in dopamine release that correlate with reduced motor activity.
Nigrostriatal overabundance of α-synuclein leads to decreased vesicle density and deficits in dopamine release that correlate with reduced motor activity.
Acta neuropathologica (2012-03-01)
Meret Nora Gaugler, Ozgur Genc, Wojciech Bobela, Safa Mohanna, Mustafa Taleb Ardah, Omar Mukhtar El-Agnaf, Marco Cantoni, Jean-Charles Bensadoun, Ralf Schneggenburger, Graham W Knott, Patrick Aebischer, Bernard Laurent Schneider
ABSTRACT
α-Synuclein (α-syn) is a presynaptic protein present at most nerve terminals, but its function remains largely unknown. The familial forms of Parkinson's disease associated with multiplications of the α-syn gene locus indicate that overabundance of this protein might have a detrimental effect on dopaminergic transmission. To investigate this hypothesis, we use adeno-associated viral (AAV) vectors to overexpress human α-syn in the rat substantia nigra. Moderate overexpression of either wild-type (WT) or A30P α-syn differs in the motor phenotypes induced, with only the WT form generating hemiparkinsonian impairments. Wild-type α-syn causes a reduction of dopamine release in the striatum that exceeds the loss of dopaminergic neurons, axonal fibers, and the reduction in total dopamine. At the ultrastructural level, the reduced dopamine release corresponds to a decreased density of dopaminergic vesicles and synaptic contacts in striatal terminals. Interestingly, the membrane-binding-deficient A30P mutant does neither notably reduce dopamine release nor it cause ultrastructural changes in dopaminergic axons, showing that α-syn's membrane-binding properties are critically involved in the presynaptic defects. To further determine if the affinity of the protein for membranes determines the extent of motor defects, we compare three forms of α-syn in conditions leading to pronounced degeneration. While membrane-binding α-syns (wild-type and A53T) induce severe motor impairments, an N-terminal deleted form with attenuated affinity for membranes is inefficient in inducing motor defects. Overall, these results demonstrate that α-syn overabundance is detrimental to dopamine neurotransmission at early stages of the degeneration of nigrostriatal dopaminergic axons.