Merck
  • The role of ozone and influence of band structure in WO3 photocatalysis and ozone integrated process for pharmaceutical wastewater treatment.
The role of ozone and influence of band structure in WO3 photocatalysis and ozone integrated process for pharmaceutical wastewater treatment.
Journal of hazardous materials (2018-08-26)
Jin Yang, Jiadong Xiao, Hongbin Cao, Zhuang Guo, Jabor Rabeah, Angelika Brückner, Yongbing Xie
ABSTRACT
Photocatalytic ozonation has great potential in wastewater treatment. However, the role of ozone and the contribution of photogenerated hole in this process have not been fully understood. Here three WO3 materials are synthesized and used as model catalysts in visible-light photocatalytic ozonation for the mineralization of pharmaceutical pollutants. A dual role of ozone in this process has been confirmed: (i) direct oxidation of the pollutant till formation of refractory intermediates, (ii) efficient trapping of photoelectron that cannot be captured by O2. The latter is crucial because it not only induces the O3--mediated pathway for hydroxyl radical (OH) formation but also separates the hole which has proven to be capable of oxidizing water into OH. Evidenced by photoluminescence results, the intrinsic charge separation ability of WO3 in photocatalytic ozonation is no more as important as that in photocatalysis with O2. Finally, this process is more applicable under acidic condition. This work contributes to a better understanding of the significance of ozone in WO3 photocatalytic ozonation and provides us an insight into the mechanism of photocatalytic ozonation.
Product Description
Cephalexin monohydrate, European Pharmacopoeia (EP) Reference Standard