Merck
  • Novel self-etching and antibacterial orthodontic adhesive containing dimethylaminohexadecyl methacrylate to inhibit enamel demineralization.
Novel self-etching and antibacterial orthodontic adhesive containing dimethylaminohexadecyl methacrylate to inhibit enamel demineralization.
Dental materials journal (2018-07-13)
Xiaomeng Wang, Ning Zhang, Bo Wang, Soo Ro Park, Michael D Weir, Hockin H K Xu, Yuxing Bai
ABSTRACT
Enamel demineralization is one of the most undesired side effects of fixed orthodontic treatment, which will lead to white spot lesions (WSLs) on tooth surfaces. The development of WSLs is due to prolonged accumulation of bacterial plaque and associated acid production. Self-etching adhesives have been used in orthodontic treatments with several advantages over the more traditional acid-etch method. However, current self-etching adhesives in orthodontic treatments have no antibacterial activity. The objectives of this study were to develop a self-etching and antibacterial orthodontic adhesive, and to investigate its enamel bond strength and antibacterial properties. A novel quaternary ammonium monomer dimethylaminohexadecyl methacrylate (DMAHDM) was incorporated into a commercial self-etching adhesive (Adper Easy One, 3M). It showed that the 5% DMAHDM appeared to be optimal in obtaining the strongest antibacterial function without compromising the enamel bond strength both at 15 min and after 30 days of immersion plus thermal cycling.
Sigma-Aldrich
1-Bromohexadecane, 97%