TSKgel® Phenyl-5PW HPLC Column
Synonym(s):
TSKgel® Hydrophobic Interaction Chromatography (HIC) Column
Products
Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing