Quercetin 3-glucoside
Synonym(s):
Quercetin 3-β-D-glucoside, 3,3′,4′,5,7-Pentahydroxyflavone 3-β-glucoside, Isoquercitrin
Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique