Skip to Content
Merck
Progesterone VETRANAL®, analytical standard

Progesterone

Progesterone VETRANAL®, analytical standard
Synonym(s):
4-Pregnene-3,20-dione, Progesterone
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique