Proteomix® SCX-NP1.7 Strong Cation Exchange Guard Cartridge and Holder
Products
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing