![NBS 127 (sulfur and oxygen isotopes in barium sulfate) NIST® RM 8557](/deepweb/assets/sigmaaldrich/product/structures/206/281/c83c9d1e-610a-45ec-b39f-c0959d331df9/640/c83c9d1e-610a-45ec-b39f-c0959d331df9.png)
NBS 127 (sulfur and oxygen isotopes in barium sulfate)
![NBS 127 (sulfur and oxygen isotopes in barium sulfate) NIST® RM 8557](/deepweb/assets/sigmaaldrich/product/structures/206/281/c83c9d1e-610a-45ec-b39f-c0959d331df9/640/c83c9d1e-610a-45ec-b39f-c0959d331df9.png)
Synonym(s):
Barium sulfate, Baryte
Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
Products
Formula Weight
Manufacturer
Format
Quality Segment
Isotope
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing