Skip to Content
Merck
Ionomycin calcium salt from Streptomyces conglobatus powder, ≥98% (HPLC)

Ionomycin calcium salt from Streptomyces conglobatus

Ionomycin calcium salt from Streptomyces conglobatus powder, ≥98% (HPLC)
Empirical Formula (Hill Notation):
C41H70CaO9
CAS Number:
Molecular Weight:
747.07

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing