
Eprosartan Related Compound E

Synonym(s):
4-(Hydroxymethyl)benzoic acid
Linear Formula:
HOCH2C6H4CO2H
CAS Number:
Molecular Weight:
152.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing