
Eprosartan Related Compound D

Synonym(s):
4-(Bromomethyl)benzoic acid, α-Bromo-p-toluic acid
Linear Formula:
BrCH2C6H4CO2H
CAS Number:
Molecular Weight:
215.04
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing