Skip to Content
Merck
CHAPS 98% (from N), suitable for dialysis, solubility: >50% (Aqueous solutions)

CHAPS

CHAPS 98% (from N), suitable for dialysis, solubility: >50% (Aqueous solutions)
Synonym(s):
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, CHAPS
Empirical Formula (Hill Notation):
C32H58N2O7S · xH2O
CAS Number:
Molecular Weight:
614.88 (anhydrous basis)

Products

Product Category
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Technique
Showing 1-1 of 1