Skip to Content
Merck
Cleland′s Reagent ULTROL® Grade, ≥99% (titration), protective agent for SH groups

DL-Dithiothreitol

Cleland′s Reagent ULTROL® Grade, ≥99% (titration), protective agent for SH groups
Synonym(s):
threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT, DL-Dithiothreitol, Cleland’s Reagent, ULTROL® Grade
Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25

Products

Brand
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reagent Type
Technique