Skip to Content
Merck
m-Cresol 99%

m-Cresol

m-Cresol 99%
Synonym(s):
1-Hydroxy-3-methylbenzene, 3-Hydroxytoluene, 3-Methylphenol
Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing