Parteck® Functional Excipients
When you design a solid dosage form, there are many factors to consider: the active ingredient, bioavailability enhancement, release kinetics, specific dosage, packaging, shelf life, and more. Our Parteck® excipients were designed with the formulator’s challenges in mind and are particle optimized for excellent performance in formulation development and manufacture. Backed by the Emprove® program with its stringent quality control, as well as regulatory support and documentation, Parteck® excipients help to speed up your approval processes.
Flexible Tableting
- High tablet hardness at low compression forces, providing tablet robustness and rapid disintegration
- High and low API dosages enabled by the unique particle surface
- Specialized excipients for direct compression, wet and dry granulation, film coating, lubrication, and rapid disintegration
Optimized Drug Delivery
- Options for modifying release kinetics, such as orally disintegrating tablets and sustained-release solid oral formulations
- Specialized excipients for different routes of administration, including dry powder inhalation
Enhanced Solubility
- Specialized excipients that boost the efficacy of your final drug product by enhancing API solubility through the use of polyvinyl alcohol for hot melt extrusion, or a silica drug carrier with a unique surface structure
Related Product Resources
Whitepaper: API Stability in Solid Dose Formulation
– Exploring the Myth of Inert Excipients
Whitepaper: New Opportunities for Oral Sustained Release
Formulations with Polyvinyl Alcohol
Whitepaper: Considerations for Dosing Accuracy
in a New Carrier for Dry Powder Inhalation
Webinar: Continuous Manufacturing in Solid Dose
– How to Leverage Opportunities of this New Technology
Webinar: Coating with Polyvinyl Alcohol: When Coating Runs Smoothly
Webinar: Hot melt extrusion with PVA
‐ A new opportunity for challenging APIs
Webinar: Solubility Enhancement, Stability and Scalability of
Mesoporous Silica Formulations: from Lab to Production Scale
Webinar: Total cost of ownership
– how the choice of excipients makes your economics