Skip to Content
Merck
All Photos(3)

Key Documents

E9908

Sigma-Aldrich

p3XFLAG-CMV-8 Expression Vector

Shuttle vector for transient expression of secreted N-terminal 3xFLAG fusion proteins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

tag

3X FLAG tagged

grade

for molecular biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

General description

p3XFLAG-CMV-8 Expression Vector is a 4.8 kb derivative of pCMV5 used to establish expression of N-terminal 3XFLAG fusion proteins in mammalian cells. The vector encodes three adjacent FLAG epitopes (Asp-Tyr-Lys-Xaa-Xaa-Asp) upstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG M2 antibody. The third FLAG epitope includes the enterokinase recognition sequence, allowing cleavage of the 3XFLAG peptide from the purified fusion protein. The preprotrypsin leader sequence precedes the 3XFLAG sequence.

p3XFLAG-CMV-8 Expression Vector is a shuttle vector for E. coli and mammalian cells. Efficiency of replication is optimal when using an SV40 T antigen-expressing host.

p3XFLAG-CMV-7-BAP Control Plasmid is a 6.2 kb derivative of pCMV5 used for transient intracellular expression of N-terminal 3XFLAG bacterial alkaline phosphatase fusion protein in mammalian cells. The vector encodes three adjacent FLAG epitopes (Asp-Tyr-Lys-Xaa-Xaa-Asp) upstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG M2 antibody. The third FLAG epitope includes the enterokinase recognition sequence, allowing cleavage of the 3XFLAG peptide from the purified fusion protein.

Vector Maps and Sequences

Components

  • p3XFLAG-CMV-8 Expression Vector 20 μg (E4151) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • p3XFLAG-CMV-7-BAP Control Plasmid 20 μg (C7472) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the human cytomegalovirus drives transcription of FLAG® fusion constructs.

Legal Information

This product is covered by the following patents owned by Sigma-Aldrich Co. LLC: US6,379,903, US7,094,548, JP4405125,EP1220933, CA2386471 and AU774216.
3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
p3xFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee Kyoung Joo et al.
Free radical biology & medicine, 139, 16-23 (2019-05-18)
Apurinic/apyrimidinic endonuclease/redox factor-1 (Ref-1), a multifunctional protein secreted from stimulated cells, has been identified as a new serological biomarker. Despite recent reports on the role of Ref-1 in inflammation, the biological function of secreted Ref-1 remains unknown, especially in vivo.
Luis R Hernández-Miranda et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(16), 6174-6187 (2011-04-22)
Cortical interneurons, generated predominantly in the medial ganglionic eminence, migrate around and avoid the developing striatum in the subpallium en route to the cortex. This is attributable to the chemorepulsive cues of class 3 semaphorins expressed in the striatal mantle
Zhiguang Wu et al.
Developmental and comparative immunology, 33(1), 97-104 (2008-09-02)
A cDNA encoding the chicken homologue of the human myelomonocytic differentiation antigen, CD14, was cloned by RT-PCR from chicken bone marrow cell RNA, using oligonucleotide primers based on the predicted cDNA sequence. The cloned chicken CD14 (chCD14) cDNA encodes an
Kazuhisa Watanabe et al.
PloS one, 8(6), e67234-e67234 (2013-07-05)
Ildr2, a modifier of diabetes susceptibility in obese mice, is expressed in most organs, including islets and hypothalamus, with reduced levels in livers of diabetes-susceptible B6.DBA mice congenic for a 1.8 Mb interval of Chromosome 1. In hepatoma and neuronal
R M Miceli et al.
Journal of immunological methods, 167(1-2), 279-287 (1994-01-03)
Libraries of random peptides can be screened to identify species which interact with antibodies or receptors. Similarly, maps of native molecular interactions can frequently be deduced by screening a limited set of peptide fragments derived from sequences within a native

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service