- IL-23 Activated γδ T Cells Affect Th17 Cells and Regulatory T Cells by Secreting IL-21 in Children with Primary Nephrotic Syndrome.
IL-23 Activated γδ T Cells Affect Th17 Cells and Regulatory T Cells by Secreting IL-21 in Children with Primary Nephrotic Syndrome.
Scandinavian journal of immunology (2017-11-10)
L Zhang, J Yan, B Yang, G Zhang, M Wang, S Dong, W Liu, H Yang, Q Li
PMID29119640
ABSTRACT
This study (1) analysed the percentage of γδ T cells, γδ T cell subsets, Th17 cells and regulatory T cells (Treg cells) and (2) determined the role of IL-23 in primary nephrotic syndrome (PNS) patients with active disease and in remission. Eighty-four patients with PNS and 51 healthy age-matched controls were included in this study. The percentage of γδ T cells, γδ T cell subsets, Th17 cells and Treg cells in peripheral blood mononuclear cells (PBMCs) were analysed by fluorescence-activated cell sorting. PMBCs from PNS patients with active disease were cultured in the presence of IL-23, IL-23 and an IL-23 antagonist, or IL23 and an anti-IL-21 monoclonal antibody (mAb). The percentage of γδ T cells, IL-23R