Merck
  • Broadening the Aldolase Catalytic Antibody Repertoire by Combining Reactive Immunization and Transition State Theory: New Enantio- and Diastereoselectivities.
Broadening the Aldolase Catalytic Antibody Repertoire by Combining Reactive Immunization and Transition State Theory: New Enantio- and Diastereoselectivities.
Angewandte Chemie (International ed. in English) (2000-01-29)
Zhong, Lerner, Barbas
ABSTRACT
Nine efficient aldolase antibodies were generated by using hapten 1. This hapten unites reactive immunization and the transition state analogue approach in a single molecule. Characterization of two of these antibodies reveals that they are highly proficient (up to 1000-fold better than any other antibody catalyst) and enantioselective catalysts for aldol and retro-aldol reactions and exhibit enantio- and diastereoselectivities opposite to that of antibody 38C2.