Merck
  • Knockdown of ZNF403 inhibits cell proliferation and induces G2/M arrest by modulating cell-cycle mediators.
Knockdown of ZNF403 inhibits cell proliferation and induces G2/M arrest by modulating cell-cycle mediators.
Molecular and cellular biochemistry (2012-02-22)
Rui Guan, Xiao-Yan Wen, Jing Wu, Rongqi Duan, Huibi Cao, Simon Lam, Defu Hou, Youdong Wang, Jim Hu, Zhuchu Chen
ABSTRACT
ZNF403, also known as GGNBP2 (gametogenetin binding protein 2), is a highly conserved gene implicated in spermatogenesis. However, the exact biological function of ZNF403 is not clear. In this study, we identified the role of ZNF403 in cell proliferation and cell-cycle regulation by utilizing short hairpin RNA (shRNA)-mediated knockdown. ZNF403-specific shRNA expressing helper-dependent adenoviral vector (HD-Ad-ZNF403-shRNA) was constructed and transduced human cell lines. ZNF403 mRNA and protein expression levels were inhibited as evidenced by real-time PCR and western blot analyses. Noticeably, we found that knockdown of ZNF403 expression suppressed cell proliferation compared to the non-target shRNA and vector controls. Furthermore, cell-cycle analysis demonstrated that downregulation of ZNF403 promoted G2/M cell-cycle arrest in a dose-dependent manner. Moreover, human cell-cycle real-time PCR array revealed that ZNF403 knockdown influenced the expression profile of genes in cell-cycle regulation. Among these genes, western blot analysis confirmed the protein up-regulation of p21 and down-regulation of MCM2 in response to the ZNF403 knockdown. Additionally, knockdown of ZNF403 also showed an anti-carcinogenetic effect on anchorage-independent growth by colony formation assay and tumor cell migration by wound-healing assay with human laryngeal cancer cell line Hep-2 cells. Altogether, our findings suggest an essential role of ZNF403 in cell proliferation and provide a new insight into the function of ZNF403 in regulating the G2/M cell-cycle transition.