Merck
  • Aberrant proteomic expression of NSRP70 and its clinical implications and connection to the transcriptional level in adult acute leukemia.
Aberrant proteomic expression of NSRP70 and its clinical implications and connection to the transcriptional level in adult acute leukemia.
Leukemia research (2014-09-02)
Dan-Bee Choi, Moo-Rim Park, Hak-Ryul Kim, Chang-Duk Jun, Hyeoung-Joon Kim, Hyeok Shim, Young-Dae Kim, Chan Choi, Keum-Ha Choi, Ki Jung Yun, Soo-Cheon Chae, Raekil Park, Seong-Kyu Choe, Young-Jin Lee, Do-Sim Park
ABSTRACT
We investigated three splicing factor proteins (SFPs; NSRP70, SRSF1, and HNRNPA1) in 187 adults with and without acute leukemia (AL). We showed that NSRP70 is a novel lymphoblastic AL (ALL) surrogate marker, which presented excellent diagnostic accuracy (92%) and disappeared during remission. Its highest molecular weight form, but not total amount, was associated with adverse genetic abnormalities in myeloid AL (AML). Furthermore, we identified that these SFPs were more prevalent in ALL than in AML; were not correlated with their mRNA levels; and their formations in AL may occur without coding mutations and relate to post-translational modifications.