Merck
  • The Egr-1 transcription factor directly activates PTEN during irradiation-induced signalling.
The Egr-1 transcription factor directly activates PTEN during irradiation-induced signalling.
Nature cell biology (2002-01-10)
T Virolle, E D Adamson, V Baron, D Birle, D Mercola, T Mustelin, I de Belle
ABSTRACT
The PTEN tumour suppressor and pro-apoptotic gene is frequently mutated in human cancers. We show that PTEN transcription is upregulated by Egr-1 after irradiation in wild-type, but not egr-1-/-, mice in vivo. We found that Egr-1 specifically binds to the PTEN 5' untranslated region, which contains a functional GCGGCGGCG Egr-1-binding site. Inducing Egr-1 by exposing cells to ultraviolet light upregulates expression of PTEN messenger RNA and protein, and leads to apoptosis. egr-1-/- cells, which cannot upregulate PTEN expression after irradiation, are resistant to ultraviolet-light-induced apoptosis. Therefore, Egr-1 can directly regulate PTEN, triggering the initial step in this apoptotic pathway. Loss of Egr-1 expression, which often occurs in human cancers, could deregulate the PTEN gene and contribute to the radiation resistance of some cancer cells.