Merck
  • Dechlorination of pentachloroethane by commercial Fe and ferruginous smectite.
Dechlorination of pentachloroethane by commercial Fe and ferruginous smectite.
Chemosphere (2002-07-11)
Javiera Cervini-Silva, Richard A Larson, Jun Wu, Joseph W Stucki
ABSTRACT
Short-term experiments were conducted to investigate the effect of a commercial Fe and an iron-bearing clay mineral, ferruginuous smectite (SWa-1), on the degradation of pentachloroethane (PCA). After 3 h of contact time, SWa-1 catalyzed PCA dehydrochlorination to tetrachloroethene (PCE, 65% conversion), whereas commercial Fe promoted PCA stepwise dechlorination via dehydrochlorination (approximately 40% conversion) and subsequent PCE hydrogenolysis to trichloroethene (TCE). The addition of unaltered SWa-1 to commercial Fe led to a complete inhibition on TCE production, whereas the addition of reduced SWa-1 barely resulted in a 30% decrease.
Sigma-Aldrich
Pentachloroethane, 95%