Merck
  • 3 beta-Hydroxysteroid dehydrogenase of Ruminococcus sp. from human intestinal bacteria.
3 beta-Hydroxysteroid dehydrogenase of Ruminococcus sp. from human intestinal bacteria.
Journal of biochemistry (1986-05-01)
T Akao, T Akao, M Hattori, T Namba, K Kobashi
ABSTRACT
Ruminococcus sp. PO1-3 obtained from human intestinal flora is able to reduce dehydrocholate as well as 3-ketoglycyrrhetinate. From this bacterium dehydrocholate- and 3-ketoglycyrrhetinate-reducing activities were purified one thousand-fold together with 3-ketocholanate-reducing and 3-beta-hydroxyglycyrrhetinate (glycyrrhetic acid) oxidizing activities by means of Matrex Red A, Sephadex G-200 and Octyl-Sepharose column chromatography. The purified enzyme catalyzed the reduction of dehydrocholic acid to 3 beta-hydroxy-7,12-diketocholanic acid and of 3-ketocholanic acid to 3 beta-hydroxycholanic acid. Studies on substrate specificity revealed that the enzyme had absolute specificity for the beta-configuration of a hydroxyl group at the 3 position of bile acid and steroids having no double bond in the A/B ring. This enzyme was neither beta-hydroxysteroid dehydrogenase [EC 1.1.1.51] nor 3 beta-hydroxy-delta 5-steroid dehydrogenase [EC 1.1.1.145], but a novel type of enzyme, defined as 3 beta-hydroxysteroid dehydrogenase.