Merck
  • Loss of Diacylglycerol Kinase-Ζ Inhibits Cell Proliferation and Survival in Human Gliomas.
Loss of Diacylglycerol Kinase-Ζ Inhibits Cell Proliferation and Survival in Human Gliomas.
Molecular neurobiology (2015-10-11)
Jinfu Diao, Chunyong Wu, Junying Zhang, Jialin Liu, Xinwu Zhang, Pengcheng Hao, Shanmin Zhao, Zhiwen Zhang
ABSTRACT
Diacylglycerol kinases ζ (DGKζ) is a critical lipid kinase which is involved in phosphatidic acid (PA) generation via diacylglycerol (DAG) phosphorylation. DGKζ is highly expressed in central nervous system and essential for brain development. Studies have indicated that DGKζ is associated with colon cancer invasion and metastasis. However, the involvement of DGKζ in human glioma development remains elusive. Here, we explored the impact and possible mechanisms of DGKζ knockdown on the proliferation and survival of glioma cells. The relationship between DGKζ expression status and human glioma stages was explored in 111 specimens of human gliomas via immunohistochemistry technology. Then the impact of DGKζ on cell proliferation, cell cycle, survival, and colony formation ability was determined in U-87 MG glioma cell lines via lentiviral-mediated small interfering (shRNA) strategy. The influence of DGKζ knockdown on global gene expression in U-87 MGglioma cell lines was further analyzed by microarray platform to reveal the possible molecular mechanisms underlying DGKζ-mediated glioma development and progression. Immunohistochemistry analysis revealed that DGKζ expression is positively correlated with human gliomagrade. Lentiviral-mediated small interfering (shRNA) strategyefficiently reduced DGKζ expression and DGKζ knockdown impaired cell proliferation, inhibited colony formation ability, and induced cell cycle arrest and cell apoptosis in U-87 MG glioma cells. Finally, microarray analysis revealed that multiple cancer-associated pathways and oncogenes were regulated by DGKζ knockdown, which provides insights into underlying mechansims of DGKζ-associated glioma development and progression. Our results established the positive correlation between DGKζ expression and gliomagrade. Furthermore, DGKζ knockdown in human glioma cell lines U-87 MG impaired cell proliferation, inhibited colony formation ability, and induced cell cycle arrest and apoptosis which microarray analysis showed that DGKζ knockdown interrupted multiple oncogenes and cancer-associated pathways. Taken together, we provided confidential evidence for the causal role of DGKζ in glioma development and progression.