Merck
  • Inhibitory effect of CP-25 on intimal formation and vascular hyperplasia via suppression of GRK2/ERK1/2/EVI1 signaling.
Inhibitory effect of CP-25 on intimal formation and vascular hyperplasia via suppression of GRK2/ERK1/2/EVI1 signaling.
Archives of biochemistry and biophysics (2020-09-28)
Jing Zhang, Yang Liu, Ming Long, Jun Li, Weikun Zhao, Qiang Su
ABSTRACT
Excessive proliferation, migration and dedifferentiation of vascular smooth muscle cells (VSMCs) are the center of intimal formation during in-stent restenosis and vein graft disease. Paeoniflorin-6'-O-benzene sulfonate (CP-25) is known to suppress inflammation and atherogenesis. However, the potential effect of CP-25 on intimal formation remains elusive. In the present study, we found that CP-25 significantly attenuated wire injury-induced intimal formation in C57BL/6 mice (intimal area: 2.64 ± 0.25 × 104 μm2 vs. 1.53 ± 0.21 × 104 μm2, P < 0.05) and vascular hyperplasia indicated by PCNA staining. In vitro experiments showed that CP-25 significantly alleviated human aortic smooth muscle cell (HASMC) proliferation, migration and dedifferentiation induced by PDGF-BB. Mechanistically, CP-25 inhibited GRK2 phosphorylation through PDGF receptor in the presence of PDGF-BB. In accordance with these results, CP-25 disrupted the interaction of GRK2 with ERK1/2 and suppressed the activation of ERK1/2 signaling in HASMCs. EVI1, which is considered as a downstream of ERK1/2 signaling and a novel transcription factor for VSMC differentiation, was also downregulated by CP-25 treatment. Moreover, overexpression of EVI1 partly restored the decreased proliferation and dedifferentiation of HASMCs treated by CP-25. Collectively, these findings suggested that CP-25 could alleviate intimal formation in response to wire injury via suppression of the interaction of GRK2 and ERK1/2 and EVI1 activation, indicating CP-25 might serve as a potent pharmaceutical for intimal formation.