Merck
  • Convenient synthesis of photoaffinity probes and evaluation of their labeling abilities.
Convenient synthesis of photoaffinity probes and evaluation of their labeling abilities.
Organic letters (2007-05-08)
Toshiyuki Kan, Yoichi Kita, Yuichi Morohashi, Yusuke Tominari, Shinnosuke Hosoda, Taisuke Tomita, Hideaki Natsugari, Takeshi Iwatsubo, Tohru Fukuyama
ABSTRACT
Convenient synthesis of a variety of photoaffinity probes was accomplished by utilizing our Ns strategy and novel resin. The synthetic probes were evaluated via the labeling ability with the preseniline 1 C-terminal fragments, which was identified as a therapeutic target for Alzheimer's disease.
Sigma-Aldrich
Biotinamidoethyldisulfanyl(2-aminoethane) trifluoroacetic acid