- Potential role of alphav and beta1 integrins as oocyte adhesion molecules during fertilization in pigs.
Potential role of alphav and beta1 integrins as oocyte adhesion molecules during fertilization in pigs.
Integrin molecules are cell adhesion molecules that are thought to be involved in sperm-oocyte interaction in rodents and humans. The objective of this study was to evaluate whether integrin molecules were present on the surface of pig oocytes, consistent with involvement in sperm-oocyte interaction in this species. Immunocytochemistry and confocal microscopy were used to evaluate the presence of beta1, and alpha1, alpha2, alpha3, alpha4, alpha5, alpha6 and alphav integrin subunits on the plasma membrane of pig oocytes. The beta1 and alphav integrin subunits were present consistently at the surface of pig oocytes; however, the remaining alpha integrin subunits evaluated were not routinely detected. The antibodies to the beta1 and alphav integrin subunits recognized appropriately sized protein bands on western blots of partially purified oocyte plasma membrane. These two antibodies also recognized oocyte plasma membrane protein isolated from a sperm plasma membrane affinity column. Sperm plasma membrane proteins of 137 and 93 kDa appeared to be the ligands for the beta1 integrin subunit as revealed by a western sandwich blot. Antibody to an extracellular domain of the beta1 integrin subunit reduced pig sperm-oocyte binding (P < 0.05), also indicating an assisting role for a beta1 oocyte integrin subunit in sperm-oocyte interaction in pigs. These results are consistent with an alphavbeta1 pig oocyte integrin interacting with a ligand on the sperm plasma membrane during fertilization.