Skip to Content
Merck
vibrant-m

SUPELCOSIL ABZ+Plus Supelguard Cartridge

vibrant-m
EC Number:

Products

Product Category
Brand
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Column Material
Particle Size (µm)
Pore Size (Å)
Technique